VH05045
Alkali Scientific™
Alkali Scientific™ CellPro™ 50mL Bottle Top Filter for 50mL centrifuge tube
PES Membrane, 0.45um, single wrapped, 24/Cs
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
Alkali Scientific™
General description
Individually Wrapped
Gamma Sterilized
Pyrogen Free
Engraved Graduation Marks
Low Protein Binding
available in 0.22µm and 0.45µm
Extremely high flow rates at very low differential pressure
Lowest Protein binding material for high recovery rates
PES (Polythersulfone) material is proven to be lower protein binding than CA (Calcium Acetate) with far superior durability
suitable for filtration of Media & other production/cleanroom tasks
filter liquid pharmaceuticals & protein solutions during medicinal drug processing
Legal Information
CellPro™ is a trademark of Alkali Scientific.
Choose from one of the most recent versions:
Certificates of Analysis (COA)