Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1061

CELLTREAT

VistaLab Pipettes

1-5mL Pipette, MLA, Macro, Adjustable

Synonym(s):

Pipettors

Sign Into View Organizational & Contract Pricing


About This Item

PDF Icon
Shipped & Sold By CELLTREAT
A Trusted Marketplace Seller
PDF Icon
Have a question?

manufacturer/tradename

CELLTREAT

General description

  • MLA® Pipettes have been a laboratory tradition for over 4 decades.
  • Made of precision engineered aluminum alloy and stainless steel, with a modern, sleek, anodized metal finish.
  • Dependable performance and durability designed for highly accurate delivery.
  • Color coded body for easy volume identification and individually serialized with Calibration Certificate included.
  • Available as single-stroke "to deliver" or two-stroke "to contain" operation.
  • Available in fixed volumes, ranging from 2µL-10mL, or adjustable volumes, ranging from 10µL-5mL.
  • MLA D-Tipper, MLA Micro D-Tipper, and MLA Digital include built-in tip ejector.
  • Limited Lifetime Warranty (covering parts).

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library