Key Documents
CH0204C
Eisco Labs
Eisco Labs Bristle Cleaning Brush
Nylon Cleaning Brush, 9.25" - Fan Shaped End - 0.5" Diameter
Synonym(s):
Brush, Glass Cleaner, Bristle Brush
About This Item
PDF Icon
Shipped & Sold By Eisco Labs
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Eisco Labs
General description
  • CLEANING BRUSH || Brush designed to scrub test tubes or other narrow glassware. Brush (including handle) measures 9.25" in total length
  • STAINLESS STEEL HANDLE || Brush features a stainless steel, twisted wire handle that is resistant to corrosion. Moderate flexibility allows you to modify the handle shape if needed
  • GREAT FOR CLEANING NARROW SPACES || Excellent for scrubbing hard-to-reach, narrow spaces including test tubes, pipes, cups, bottles, jars, vases, laboratory glassware, measuring cylinders, beakers and more
  • FANNED TIP || Brush features semi-stiff, nylon fiber bristles with a fanned end for scrubbing tubes with rounded ends. Fiber section measures 3.5" in length and is 0.5" in diameter
  • DESIGNED FOR PROFESSIONAL LABORATORIES || Designed for use in professional laboratories, but is perfect for use in the home and garage
This semi-stiff, nylon fiber bristle brush is perfect for scrubbing hard to reach, narrow spaces - straws, pipes, cups, bottles, jars, vases, laboratory glassware, measuring cylinders, beakers and more. The fanned end allows you to easily clean rounded surfaces and corners. The stainless steel, twisted handle is semi-flexible, allowing you to modify the shape as needed. Looped end for easy hanging. Designed for professional science laboratories but is perfect for a variety of applications in the home or garage.
Brush measures 9.25" in total length, with the bristle section measuring 3.5" in length and 0.5" in diameter, and handle measuring 5.75". Ideal for tubes with diameters of 0.3" - 0.5".
Choose from one of the most recent versions:
Certificates of Analysis (COA)