Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CH0341DBL

Eisco Labs

Eisco Labs Class A Graduated Glass Cylinder

Graduated Glass Cylinder, 100mL - ASTM, Class A ,Hexagon Base

Synonym(s):

Glass Cylinder, Graduated Cylinder

Sign Into View Organizational & Contract Pricing


About This Item

PDF Icon
Shipped & Sold By Eisco Labs
A Trusted Marketplace Seller
PDF Icon
Have a question?

manufacturer/tradename

Eisco Labs

General description

  • INDUSTRIAL QUALITY || Made from heavy-duty borosilicate 3.3 glass.
  • EXTREMELY ACCURATE || Meets ASTM Class A tolerances for precision and are individually calibrated to contain.
  • EASY TO READ || Large, blue single scale in 1.0 mL graduations.
  • MAXIMUM STABILITY || One-piece molded glass construction with hexagonal base and adjustable plastic bumper guard.
  • GREAT FOR PROFESSIONAL LABORATORIES, CLASSROOMS and HOUSEHOLD || Superior chemical durability and heat tolerance. Designed for use in professional laboratories, but is an ideal choice for science classrooms and household use.

100mL capacity measuring cylinder made of high quality, heavy duty borosilicate 3.3 glass. Autoclavable, with superior chemical durability. Low temperature gradient - can withstand high temperatures and thermal shocks. With large, blue single scale in 1.0mL graduations and hexagonal base for extra stability. Class A tolerance, ±0.50mL as per ISO/ASTM. Fitted with a plastic, hexagonal protective collar. Cylinder stands 9.75" tall, with a base measuring 3" wide. Inner diameter of cylinder body measures 29.2mm (1.15"), and outer diameter measures 31.8mm (1.25").


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library