Key Documents
CH0354G
Eisco Labs
Eisco Labs Class B Graduated Plastic Cylinder
Graduated Plastic Cylinder, 1000mL - Class B, Octagon Base- Autoclavable
Synonym(s):
Plastic Cylinder, Graduated Cylinder
About This Item
PDF Icon
Shipped & Sold By Eisco Labs
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Eisco Labs
General description
  • INDUSTRIAL QUALITY || 1000ml capacity measuring cylinder. Class B tolerance. The translucent, rigid polymer construction is resistant to fatigue for superior and dependable durability
  • OCTAGONAL BASE || Cylinder features a octagonal base for maximum stability
  • RAISED GRADUATIONS || Superior optical clarity with easy to read, raised 10ml graduation lines that are marked every 100ml. Calibrated to contain
  • POLYPROPYLENE CONSTRUCTION || Made of high quality polypropylene that features high heat tolerance, and good to excellent chemical resistance. Autoclavable for easy cleaning and sterilization
  • GREAT FOR PROFESSIONAL LABORATORIES, CLASSROOMS and HOUSEHOLD || Designed for use in professional laboratories, but is an ideal choice for science classrooms and household use
Measuring cylinder with a 1000ml capacity made of premium quality polypropylene plastic. Class B tolerance. Calibrated to contain.
Translucent rigid construction and superior optical clarity. Easy to read, raised graduation lines every 10ml, and marked every 100ml. Octagonal base for extra stability, and a spout for easy pouring.
Microwavable and resistant to fatigue for long term durability. High heat tolerance with a temperature range of -20 to +135°C. Good to excellent chemical resistance. Autoclavable at 121°C for easy cleaning and sterilization.
Cylinder stands 16" tall, with a base measuring 5" wide. Outer diameter of cylinder body measures 2.9", and inner diameter measures 2.7".
Choose from one of the most recent versions:
Certificates of Analysis (COA)