Key Documents
CH0390F
Eisco Labs
Eisco Labs 100pk Filter Paper
100pk Filter Paper, 21.5cm Dia., Medium Speed
Synonym(s):
Filtering, Filter, Filter Paper, Lab Paper
About This Item
PDF Icon
Shipped & Sold By Eisco Labs
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Eisco Labs
General description
  • High retentivity, perfect for general laboratory filtering and qualitative analysis
  • 10µL (micron) pore size
  • Medium Speed (85 gsm)
  • Pack of 100 circles
Premium quality 21.5cm diameter filter paper for use in labs. Medium speed. Pore size: 10µL. Pack of 100 circles.
Choose from one of the most recent versions:
Certificates of Analysis (COA)