CH0573E
Eisco Labs
Eisco Labs Class A Serological Glass Pipette with Blue graduation
Serological Pipette, 25ml - Class A, Tolerance ±0.100ml - Blue Graduations - Color Code, White - Calibrated for Delivery to Jet - Borosilicate Glass
Synonym(s):
Serological Pipette, Serological, Pipette, Glass Pipette
About This Item
Recommended Products
manufacturer/tradename
Eisco Labs
General description
- BOROSILICATE 3.3 GLASS || Serological pipette made of high quality Borosilicate 3.3 glass. Ideal for transferring liquids, mixing chemical solutions or working with reagents with varying densities. Excellent for professional laboratories, research, science classrooms or household use. Autoclavable, with superior chemical durability. Low temperature gradient- can withstand high temperatures & thermal shocks
- 25ML CAPACITY || Serological pipette with a 25ml capacity. Color code, white. Eisco Labs serological pipettes feature color coding, allowing you to easily identify the size
- CLASS A TOLERANCE || Class A tolerance ±0.100ml, as per DIN IS 835 standard. Calibrated for delivery to jet. Last drop expelled by blowing
- BLUE GRADUATIONS || Pipette features blue graduations for optimal clarity. 0.2ml graduation lines, marked every 2ml
- MANUFACTURED BY EISCO LABS || Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Serological pipette made of high quality, heavy duty Borosilicate 3.3 glass. 25ml capacity, 0.2ml blue graduation lines, marked every 2ml. Class A, tolerance, ±0.100ml . Calibrated for delivery to jet.
Pipette measures 14.25" in length, and has a body diameter of 0.6". Top has an inner diameter of 5mm and an outer diameter of 8mm.
Autoclavable, with superior chemical durability. Low temperature gradient- can withstand high temperatures & thermal shocks. Eisco Labs Borosilicate glass has the following heat tolerances: Strain Point -515°C | Annealing Point 565°C | Softening Point 820°C.
Eisco Labs proudly manufactures its own products using state-of-the-art equipment, and following the highest standards of quality control.
Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.