Key Documents
FL0705
Eisco Labs
Eisco Labs Glass Narrow Neck Erlenmeyer Flasks
12PK Glass Narrow Neck Erlenmeyer Flasks, 250mL
Synonym(s):
Narrow Neck Flasks, Glass Flasks, Erlenmeyer Flask
About This Item
PDF Icon
Shipped & Sold By Eisco Labs
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Eisco Labs
General description
  • 250mL CAPACITY || Pack of 12 Erlenmeyer Flasks, each with 250mL capacity and easy-to-read white enamel graduations every 12.5mL
  • INDUSTRIAL QUALITY || Made of high quality, heavy duty, borosilicate 3.3 glass with excellent strength against breakage. Microwavable and autoclavable, with superior chemical durability. Can withstand high temperatures and thermal shocks
  • SUPERIOR DURABILITY || Features uniform wall thickness ideal for heating and a reinforced rim for protection against high impacts
  • MEETS DIN ISO STANDARDS || Conforms with DIN ISO 1773 standards for narrow neck conical flasks
  • GREAT FOR PROFESSIONAL LABORATORIES, CLASSROOMS and HOUSEHOLD || Superior chemical durability and heat tolerance. Designed for use in professional laboratories, but is an ideal choice for science classrooms and household use
Pack of 12 250mL Erlenmeyer Flasks with narrow necks are made of heavy-duty borosilicate 3.3 glass. Features white enamal graduations every 12.5mL, a uniform wall thickness ideal for heating, and a reinforced rim to protect against accidental impacts. Meets DIN ISO 1773 standards for conical flasks.
Choose from one of the most recent versions:
Certificates of Analysis (COA)