Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1501030-FLS

Foxx Life Sciences

Foxx Life Sciences PUREGRIP® Media Bottle GL45 VersaCap, 2000mL

Glass Media Bottles, Round, Clear, GL45 VersaCap®, 2,000mL, 10/CS

Synonym(s):

Bottles, Media Bottles

Sign Into View Organizational & Contract Pricing


About This Item

PDF Icon
Shipped & Sold By Foxx Life Sciences
A Trusted Marketplace Seller
PDF Icon
Have a question?

capacity

2000 mL

feature

with cap

material

Bottle: borosilicate glass 3.3
Cap: polypropylene

manufacturer/tradename

Foxx Life Sciences

General description

Foxx PUREGRIP® Glass Reagent Bottles are chemically inert and stable. When fitted with a plastic pouring ring, dripping can be totally eliminated. As there is only one size of screw thread for all the bottles from 100 ml onwards, the screw caps and pouring rings are fully interchangeable. The bottles, pouring rings and caps are autoclavable and sterilizable.

  • Made of high quality borosilicate glass
  • Clear or Amber borosilicate glass
  • Pour Ring for Drip prevention
  • Available in 100ml, 150ml 250 ml, 500ml, 1L, 2L, 5L, 10L, and 20L capacity
  • Perfect for any science or home laboratory

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library