Key Documents
302-1101-FLS
Foxx Life Sciences
Vactrap™ 2 Vacuum Trap Bottle (Vacuum Flask Alternative)
For Aspiration and Vacuum Protection, Shatterproof, 1L + 1L, HDPE Bottle, Red Bin, 1/4" ID Tubing, 1/EA
Synonym(s):
Vacuum Trap
About This Item
PDF Icon
Shipped & Sold By Foxx Life Sciences
A Trusted Marketplace Seller
PDF Icon
Have a question?
material
bottle: high-density polyethylene
filter: null
manufacturer/tradename
Foxx Life Sciences
General description
Foxx Life Sciences' Vactrap2™ is the world's most advanced vacuum trap system. Ideal for the protection of vacuum lines and scientists alike, Vactrap2™ safely contains waste from supernatant removal, chemical separation, and cell or tissue culture media aspiration. Best of all, Vactrap2™ comes ready to use; simply attach to your vacuum and begin your work.
The full Vactrap2™ system includes one 1L + 1L Foxx Heavy Duty Vacuum Bottles (Bleach Compatable HDPE), with a patented VersaCap® closure for tangle-free opening, no need to disconnect tubing, ever. NEW Improved heavy duty EZBio® TPE V tubing to avoid kinking. A 0.2µm PTFE inline vent filter retains any aerosolized bacteria. A pinch clamp allows for modulation of vacuum. Other Vactrap2™ options offer two bottles or setups with secondary containment.
Say goodbye to glass. Say hello to Vactrap2™.
DO NOT USE CONTINUOUSLY UNDER VACUUM FOR 12 OR MORE HOURS. If a longer time is needed please purchase Vactrap™ G (Glass) https://www.foxxlifesciences.com/collections/vactrap-g
Choose from one of the most recent versions:
Certificates of Analysis (COA)