Key Documents
3039Y
Globe Scientific
Globe Scientific 1mL and 2mL Cryogenic Vial Storage Box, 25 Place, BioBox
BioBOX 25, for 1.0mL and 2.0mL CryoCLEAR vials, Polycarbonate (PC), Holds 25 vials (5x5 format), Printed Lid, Pack Includes a CryoClear Tube Picker, YELLOW-BOX/8
About This Item
PDF Icon
Shipped & Sold By Globe Scientific LLC
A Trusted Marketplace Seller
PDF Icon
Have a question?
General description
Produced from polycarbonate (PC), these storage boxes are designed to withstand temperatures of -196°C to +121°C. The boxes fit in stainless steel freezer racks and have vent holes for drainage and rapid freezing.
The lid is transparent for viewing contents and has a printed numerical grid for easy sample identification. Each box features two angled corners to guarantee proper alignment of the grid. A frosted writing area provides the perfect surface for smear-resistant marketing.
Each pack of boxes comes with a free vial picker.
The boxes are available in 25-place, 81-place, and 100-place sizes and are available in assorted colors.
Choose from one of the most recent versions:
Certificates of Analysis (COA)