Key Documents
601825-1
Globe Scientific
Globe Scientific Griffin Style Beakers, Polypropylene, Printed Graduations
Beaker, PP, Printed Graduations, 500mL-1EA
About This Item
PDF Icon
Shipped & Sold By Globe Scientific LLC
A Trusted Marketplace Seller
PDF Icon
Have a question?
General description
These superior quality Griffin beakers are ideal for all general laboratory procedures. In one beaker you can mix, measure and prepare solutions. Our Griffin beakers have a low-form and a tapered pour spout to facilitate the transferring of liquids without dripping. The heavy-duty plastic withstands repeated autoclaving. Switching from glass beakers to Globe′s plastic equivalents will greatly reduce breakage and the potential for injury.
DON'T NEED A CASE? BUY BY THE EACH!
  • Special no-drip pour spout.
  • Graduated in milliliters (mL).
  • Break-resistant
Choose from one of the most recent versions:
Certificates of Analysis (COA)