Key Documents
8111000
Globe Scientific
Globe Scientific Globe Glass Amber Media Bottles
Bottle, Amber Media, Globe Glass, 1000mL, GL45 Screw Cap, Dual Graduations, 10/Box
About This Item
PDF Icon
Shipped & Sold By Globe Scientific LLC
A Trusted Marketplace Seller
PDF Icon
Have a question?
feature
with cap
material
Bottle: borosilicate glass
General description
Globe Glass™ amber media bottles are designed with a wide mouth for ease of filling, dispensing, and pipetting. A drip-free pour ring and autoclavable polypropylene (PP) screw cap enable sterilization of media and reagents. Globe Glass amber media bottles are a great choice for mixing, storing, and transport of light-sensitive solutions and media. Available in sizes 50mL to 2000mL.
  • ASTM E438, Type I, Class A, Borosilicate 3.3 Glass
  • Amber color reduces light transmission
  • GlassGuard™ packaging system protects against transit damage
  • GL32 polypropylene (PP) screw cap on size 50mL
  • GL45 polypropylene (PP) screw caps on sizes 100mL to 2000mL
  • Pouring ring for drip-free liquid transfer
  • High resistance to chemicals
  • Dual scale metric graduations
  • Durable white enamel printing
  • Large white marking area
Choose from one of the most recent versions:
Certificates of Analysis (COA)