05-1308-AF647
Anti-Ubiquitin, Lys63 Antibody, clone Apu3, Alexa Fluor 647 Conjugate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
General description
Immunogen used was human di-ubiquitin linked at Lys63.
Specificity
Recognizes ubiquitin linked at Lys63. Does not recognize ubiquitin linked at Lys48 or free ubiquitin.
Application
Epitope: Lys63 ubiquitination site.
Quality
Evaluated by Immunocytochemistry in HeLa cells.
Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Lys63-linked ubiquitin in HeLa cells.
Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Lys63-linked ubiquitin in HeLa cells.
Physical form
Purified rabbit monoclonal IgG antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % sodium azide.
Storage and Stability
Stable for 1 year at 2-8°C from date of receipt.
Legal Information
ALEXA FLUOR is a trademark of Life Technologies
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service