Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07-1441

Sigma-Aldrich

Anti-phospho-Paxillin (Ser126) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

paxillin, FLJ16691

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer126)

Gene Information

human ... PXN(5829)
rat ... Pxn(360820)

General description

Paxillin, a focal adhesion protein, is involved in focal adhesion formation during cell adhesion and migration. Paxillin contains LD motifs, LIM domains, and SH3-/SH2-binding domains that participate in a variety of protein-protein interactions with kinases, GTPase-activating proteins, and cytoskeletal proteins. Phosphorylation of paxillin occurs at both tyrosine and serine sites. Serine phosphorylation of paxillin occurs in response to growth-factor activation and fibronectins. Both JNK1 and cdc2 kinases can phosphorylate serine 178 in paxillin. The mutant form of paxillin (S178A) decreases the migration of keratocytes and epithelial cells.

Specificity

This antibody recognizes Paxillin phosphorylated at Ser126.

Immunogen

Epitope: Ser126
KLH-conjugagted synthetic peptide corresponding to human Paxillin at phosphorylated Ser126.

Application

Anti-phospho-Paxillin (Ser126) Antibody is an antibody against phospho-Paxillin (Ser126) for use in WB.
Research Category
Signaling
Research Sub Category
G-proteins

Quality

Evaluated by Western Blot in pervandate treated and untreated Huvec cell lysate.
Western Blot Analysis: 1:1,000 dilution of this antibody detected Paxillin on 10 µg of Pervandate treated and untreated Huvec cell lysate.

Target description

68 kDa

Linkage

Replaces: AB3793

Physical form

Affinity purified
PBS (no Ca2+, no Mg2+), pH 7.3, 50% glycerol, 1 mg/mL BSA, and 0.05% sodium azide.

Storage and Stability

Maintain at -20°C for up to one year or 2-8°C for short term use from date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Pervandate treated and untreated Huvec cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service