Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.04737

Immersion oil

new

for microscopy

Sign Into View Organizational & Contract Pricing


About This Item

Quality Level

color

light yellow

refractive index

n20/D 1.515-1.517

density

1.02 g/cm3 at 20 °C

storage temp.

15-25°C

General description

Immersion Oil - for Microscopy, is of an oily nature and has a defined refractive index of about 1,516. It is important that the refractive index (nD) is about 1.5, similar to that of glass. This enables a homogeneous oil immersion to be achieved. Immersion media are used in conjunction with immersion objectivs and are located between the surface of the specimen and the lens of the microscope. Immersion oils practically eliminate light beam deflection so that the effectiveness of the lens is considerably increased. Immersion Oil - for Microscopy is produced from a natural component.

Application

diagnostic assay

manufacturing hematology

histology

Analysis Note

Refractive index (n 20/D): 1.515 - 1.517 Density (d 20 °C/ 4 °C): 1.0245 - 1.0265 Transmission (380 nm; 1 cm): ≥ 65 % Transmission (400 nm; 1 cm): ≥ 78 % Transmission (450 nm; 1 cm): ≥ 90 % Fluorescence (as quinine at 365 nm): ≤ 1500 ppb Viscosity (20 °C): 100 - 120 mPa∙s Suitability for microscopy: passes test

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service