Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-641-M

Sigma-Aldrich

DUSP22 Protein, 10 µg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

biological source

human

Quality Level

description

protein target DUSP22

purified by

(Glutathione-agarose)

suitability

suitable for phosphatase test

UniProt accession no.

Gene Information

human ... DUSP22(56940)

General description

FUNCTION: SwissProt: Q9NRW4 # Activates the Jnk signaling pathway. Dephosphorylates and deactivates p38 and stress-activated protein kinase /c-Jun N- terminal kinase (SAPK/JNK) (By similarity).
SIZE: 184 amino acids; 20910 Da
SUBCELLULAR LOCATION: Cytoplasm (By similarity). Nucleus (By similarity).
TISSUE SPECIFICITY: Ubiquitous. Highest expression seen in heart, placenta, lung, liver, kidney and pancreas.
SIMILARITY: SwissProt: Q9NRW4 ## Belongs to the protein-tyrosine phosphatase family. Non-receptor class dual specificity subfamily. & Contains 1 tyrosine-protein phosphatase domain.

Packaging

Also available in 10 μg size (cat#14-641) and in bulk (cat#14-641-K).

Quality

Routinely evaluated in a phosphatase assay

Physical properties

47.3kDa

Storage and Stability

6 months at -70°C

Analysis Note

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service