Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

324673

Sigma-Aldrich

EGFR/ErbB-2 Inhibitor

The EGFR/ErbB-2 Inhibitor, also referenced under CAS 179248-61-4, controls the biological activity of EGFR/ErbB-2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

EGFR/ErbB-2 Inhibitor, 4557W, 4-(4-Benzyloxyanilino)-6,7-dimethoxyquinazoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H21N3O3
CAS Number:
Molecular Weight:
387.43
MDL number:
UNSPSC Code:
12352202

Quality Level

assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

methanol: 1 mg/mL
DMSO: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N(c3ccc(cc3)OCc4ccccc4)c1ncnc2c1cc(c(c2)OC)OC

InChI

1S/C23H21N3O3/c1-27-21-12-19-20(13-22(21)28-2)24-15-25-23(19)26-17-8-10-18(11-9-17)29-14-16-6-4-3-5-7-16/h3-13,15H,14H2,1-2H3,(H,24,25,26)

InChI key

DNOKYISWMVFYFA-UHFFFAOYSA-N

General description

A cell-permeable 4-anilino quinazoline compound that acts a potent, reversible, and ATP-competitive inhibitors of EGFR and c-erbB2 (IC50 = 20 nM and 79 nM, respectively), members of the type I growth factor receptor family that is involved in breast cancer development and progression. Shown to effectively inhibit the proliferation (IC50 = ~ 1.2-2.5 µM) of tumor cells overexpressing EGRF or c-erbB-2.
A cell-permeable, potent, reversible, and ATP-competitive inhibitor of the tyrosine kinase activity of EGFR (IC50 = 20 nM) and c-erbB2 (IC50 = 79 nM), members of the type I growth factor receptor family implicated in the development and progression of various forms of cancer, including breast and small lung cell carcinoma. Shown to effectively inhibit the proliferation (IC50 ~1.2-2.5 µM) of tumor cells overexpressing EGRF or c-erbB-2.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
EGFR
Product competes with ATP.
Reversible: yes
Target IC50: 20 nM and 79 nM, against EGFR and c-erbB2, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Cockerill, S., et al. 2001. Bioorg. Med. Chem. Lett.11, 1401.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service