69771-M
pET-23(+) DNA - Novagen
Synonym(s):
Novagen pET System, pET-23(+) DNA, pET-23(+) Vector, pET-23(+) transcription vector
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106610
NACRES:
NA.51
manufacturer/tradename
Novagen®
Quality Level
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
shipped in
dry ice
wet ice
storage temp.
−70°C
General description
pET-23(+) is a transcription vector designed for expression from bacterial translation signals carried within a cloned insert. It therefore lacks the ribosome binding site and ATG start codon present on the pET translation vectors. A C-terminal His-Tag sequence is available. Note that the sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map (TB064).
The pET Vectors are supplied as purified plasmid DNA (10 µg). Each order of pET DNA also includes an Induction Control strain (supplied as a glycerol stock). Please contact technical service if you need additional information.
The pET Vectors are supplied as purified plasmid DNA (10 µg). Each order of pET DNA also includes an Induction Control strain (supplied as a glycerol stock). Please contact technical service if you need additional information.
Application
pET-23(+) is a transcription vector that lacks the ribosome binding site and ATG start codon present on the pET translation vectors.
Warning
Toxicity: Standard Handling
Legal Information
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
This product is sold for internal research use only. Any commercial use of this product, its components, and/or any derivatives thereof (including but not limited to proteins produced using the product or its components) (together and hereinafter the ′EMD Product′) requires signature of a written commercial use agreement with EMD Millipore Corporation or its successor-in-interest. Commercial use shall include but not be limited to: (1) use of the EMD Product to manufacture products for sale to third parties; (2) use of the EMD Product to provide services, information, or data to third parties in exchange for consideration; (3) use of the EMD Product for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the EMD Product, whether or not such EMD Product is resold for research use. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of the EMD Product. Please direct any questions on these use restrictions to: licensing@milliporesigma.com.
This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges MilliporeSigma to request certain information about you and the establishment where the GMOs are being handled. Clickhere for Enduser Declaration (EUD) Form
This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges MilliporeSigma to request certain information about you and the establishment where the GMOs are being handled. Clickhere for Enduser Declaration (EUD) Form
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service