Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.20581

SAFC

Bovine Transferrin Iron Poor (APO)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
eCl@ss:
32011401

biological source

bovine

Quality Level

shelf life

48 mo.

storage temp.

-10 to -25°C

General description

Product Source: Bovine; Adult Bovine Serum

Storage and Stability

refrigerated

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service