Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB22

Sigma-Aldrich

Goat Anti-Human IgG (Heavy and Light Chain) (1L)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

antibody form

serum

Quality Level

clone

polyclonal

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable

Physical form

0.01M PBS containing 0.05% Sodium Azide as a preservative. Nonsterile liquid filtered through a 0.45 micron filter.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to one year. Avoid repeated freeze/thaw cycles.

Disclaimer

This product is intended for use by IVD manufacturers.
Contact customer service for assistance: oem.reagents@emdmillipore.com
Minimum order quantity required to qualify. Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service