Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB5861

Sigma-Aldrich

Anti-Melanin Concentrating Hormone Receptor Type 2 Antibody, intracellular, CT

Chemicon®, from chicken

Synonym(s):

MCH-R2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

chicken

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... MCHR2(84539)

General description

Melanin Concentrating Hormone Receptor type 2 (MCH-R2) is a G-protein coupled receptor with seven transmembrane domains. MCH-R2 is 340 amino acids in length (approximately 39 kDa) and shares approximately 38% amino acid identity with MCH-R1. Like MCH-R1, it binds MCH with high affinity. However, reports indicate that signaling through this receptor is pertussis insensitive, contrary to MCH-R1. Expression of MCH-R2 mRNA is restricted to several regions of the brain, including the arcuate nucleus and the ventral medial hypothalamous (Sailer, 2001).

Specificity

Human Melanin Concentrating Hormone Receptor Type 2 (MCH-R2). The antibody has been used to stain tissues from rat and mouse as well as human.

Immunogen

Epitope: intracellular, C-terminus
Synthetic 16 amino acid peptide corresponding to the intracellular carboxy-terminus of human MHC-R2.

Application

Immunohistochemistry: 1 μg/mL on brain cryostat sections. The antibody stains extensively through the brain and is particularly striking in the ganglionic layer of the cerebellum.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
CNS Control of Metabolism

Hormones & Receptors
This Anti-Melanin Concentrating Hormone Receptor Type 2 Antibody, intracellular, C-terminus is validated for use in IH for the detection of Melanin Concentrating Hormone Receptor Type 2.

Physical form

Affinity purified immunoglobulin in PBS, pH 7.2 containing 0.1% sodium azide.

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 6 months after date of receipt. The antibody can also be diluted 1:1 with glycerol (ACS grade or better) for added stability. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service