Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

ABE2695

Sigma-Aldrich

Anti-eIF-4E

from rabbit, purified by affinity chromatography

Synonym(s):

Eukaryotic translation initiation factor 4E, eIF4E, eIF-4F 25 kDa subunit, mRNA cap-binding protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, human

species reactivity (predicted by homology)

monkey (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... EIF4E(1977)

General description

Eukaryotic translation initiation factor 4E (UniProt: P06730; also known as eIF-4E, eIF4E, eIF-4F 25 kDa subunit, mRNA cap-binding protein) is encoded by the EIF4E (also known as EIF4EL1, EIF4F) gene (Gene ID: 1977) in human. elF-4E is a cytoplasmic factor that recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation of protein synthesis and facilitates ribosome binding by inducing the unwinding of the mRNAs secondary structures. It is a component of the CYFIP1-EIF4E-FMR1 complex that binds to the mRNA cap and mediates translational repression. In the CYFIP1-EIF4E-FMR1 complex. Three isoforms of elf-4E have been described that are generated by alternative splicing. Elf-4E contains four 7-methylguanosine-containing mRNA cap binding regions and three EIF4EBP1/2/3 binding regions. Hypophosphorylated EIF4EBP1, EIF4EBP2, and EIF4EBP3 are shown to compete with EIF4G1/EIF4G3 to interact with elF-4E. Phosphorylation of ElF4EBP1 by insulin-stimulated MAP kinase (MAPK1 and MAPK3) cause dissociation of the complex and allows EIF4G1/EIF4G3 to bind and result in initiation of translation. Mutations in ElF4E gene have been linked to autism with impairments in social interaction and communication and developmental abnormalities.

Specificity

This rabbit polyclonal antibody detects Eukaryotic translation initiation factor 4E in Human, Mouse, and Rat cells. it targets an epitope with in 14 amino acids from the N-terminal region.

Immunogen

Epitope: unknown
KLH-conjugated linear peptide corresponding to 14 amino acids from the N-terminal region of human Eukaryotic translation initiation factor 4E.

Application

Anti-eIF-4E, Cat. No. ABE2695, is a highly specific rabbit polyclonal antibody that targets Eukaryotic translation initiation factor 4E and has been tested in Immunocytochemistry and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: 0.9 µg/mL from a representative lot detected eIF-4E in 10 µg of PC12 cell lysate.

Immunocytochemistry Analysis: A 1:100-500 dilution from a representative lot detected eIF-4E in HeLa, A431, HUVEC, and NIH/3T3 cell lines.

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected eIF-4E in 10 µg of Jurkat cell lysate.

Target description

~31 kDa observed; 25.10 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service