Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABN168

Sigma-Aldrich

Anti-CD6 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

T-cell differentiation antigen, T-cell differentiation antigen CD6, CD6, T12, TP120

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CD6(923)

General description

T-cell differentiation antigen CD6, also known as CD_antigen CD6, T12, and TP120. CD6 is a monomeric 105- or 130-kD membrane glycoprotein that is involved in T-cell activation. The size difference between the two CD6 forms is due to differences in phosphorylation. CD6 mediates cellular adhesion and migration across the endothelial and epithelial cells. CD6 may bind in trans to surface glycoproteins. These transinteractions are reinforced during autoimmune diseases, like rheumatoid arthritis (RA), Sjögren′s syndrome, and multiple sclerosis and some cancers.

Immunogen

KLH-conjugated linear peptide corresponding to human CD6.

Application

Anti-CD6 Antibody is a highly specific rabbit polyclonal antibody, that targets CD6 & has been tested in IHC.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Immunohistochemistry in human spleen tissue.

Imunnohistochemistry Analysis: A 1:100 dilution of this antibody detected CD6 in human spleen tissue.

Target description

140 kDa calculated

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human spleen tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service