ABS1679M
Anti-VPS28
from rabbit
Synonym(s):
Vacuolar protein sorting-associated protein 28 homolog, H-Vps28, ESCRT-I complex subunit VPS28
About This Item
biological source
rabbit
Quality Level
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human
technique(s)
immunofluorescence: suitable
western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... VPS28(51160)
General description
Specificity
Immunogen
Application
Signaling
Immunofluorescence Analysis: A representative lot detected VPS28 in HEK293T cells transfected with VPS28 (Silvestri, L.S., et. al. (2007). J Infect Dis. 196 Suppl 2:S264-70).
Quality
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected recombinant GST-VPS28 protein.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service