Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABS381

Sigma-Aldrich

Anti-GBP5 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Guanylate-binding protein 5, GBP-TA antigen, GTP-binding protein 5, GBP-5, Guanine nucleotide-binding protein 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GBP5(115362)

General description

GBP5, also known as GBP-TA antigen, or GTP-binding protein 5, or Guanine nucleotide-binding protein 5 or GBP-5, and encoded by the gene GBP5, is a protein that plays a role in the inflammatory response to pathogens and may also play a role in apoptosis. GBP5 stimulates cryopryrin (NLRP3 protein) assembly of the intracellular inflammasome, a multiprotein oligomer consisting of caspase 1, PYCARD, NALP and sometimes caspase 5. Inflammasomes act as sensory complexes that alert the immune system to the presence of infection or tissue damage. Nucleosomes and GBP5 are expressed in myeloid cells and are a component of the innate immune system; GBP5 acts as key rheostat for activation and assembly of the inflammasomes. Additionally, GBP5 is a GTPase that homodimerizes upon GTP-binding; dimerization is required for the second hydrolysis step from GDP to GMP. GBP5 is localized to the cytoplasm and intracellular membranes via its lipid-anchor sequences.

Immunogen

Linear peptide corresponding to human GBP5.

Application

Detect GTP binding protein using this rabbit polyclonal antibody, Anti-GBP5 Antibody validated for use in western blotting.
Research Category
Signaling
Research Sub Category
Immunological Signaling
Western Blotting Analysis: A representative lot from an independent laboratory detected GBP5 in differentiating THP-1 cells which were untreated or treated with LPS alone, or LPS/IFN-gamma (Shenoy, A. R., et al. (2012). Science. 336(6080):481-485.).

Quality

Evaluated by Western Blotting in K562 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected GBP5 in 10 µg of K562 cell lysate.

Target description

~67 kDa observed. A second isoform is known to exist at 55 kDa Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Avinash R Shenoy et al.
Science (New York, N.Y.), 336(6080), 481-485 (2012-03-31)
Inflammasomes are sensory complexes that alert the immune system to the presence of infection or tissue damage. These complexes assemble NLR (nucleotide binding and oligomerization, leucine-rich repeat) or ALR (absent in melanoma 2-like receptor) proteins to activate caspase-1 cleavage and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service