Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABS434

Sigma-Aldrich

Anti-ApoA4 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Apolipoprotein A4, Apolipoprotein A-IV, ApoA-IV, APO-AIV, ApoA4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... APOA4(337)

General description

ApoA4, also known as Apolipoprotein A4 or ApoA-IV, is a member of the apolipoprotein A1/A4/E family. ApoA4 is a major component of HDL and it can be found on the surface of newly synthesized chylomicrons. Studies have shown that APoA4 is required for efficient activation of lipoprotein lipase by ApoC-II, which is a potent activator of Lecithin Cholesterol Acyltransferase (LCAT). Other evidence points out that ApoA4 may have a role in chylomicrons and VLDL secretion and catabolism. The physiological function of ApoA4 is not confirmed yet, but evidence from several in vitro studies indicate that it may play a role in the reverse cholesterol transport pathway, transporting cholesterol from peripheral cells back to the liver. ApoA4 is synthesized primarily in the intestine and secreted in the plasma.

Immunogen

KLH-conjugated linear peptide corresponding to human ApoA4.

Application

Anti-ApoA4 Antibody is a highly specific rabbit polyclonal antibody, that targets Apolipoprotein A & has been tested in western blotting & IHC.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected ApoA4 in human colon tissue.
Research Category
Signaling
Research Sub Category
Lipid Metabolism & Weight Regulation

Quality

Evaluated by Western Blotting in human liver tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected ApoA4 in 15 µg of human liver tissue lysate.

Target description

~38 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human liver tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service