Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AP156-K

Sigma-Aldrich

Goat Anti-Rabbit IgG (Fc) Affinity Purified (1mg)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

antibody form

affinity purified immunoglobulin

Quality Level

Physical form

Liquid. Buffer: 0.01M Tris-HCl, 0.25M NaCl, pH 8.0 with no preservatives

Storage and Stability

Refrigerator (2–8°C)

Disclaimer

This product is intended for use by IVD manufacturers.
Contact customer service for assistance: oem.reagents@emdmillipore.com
Minimum order quantity required to qualify. Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service