Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

FCMAB179F

Sigma-Aldrich

Milli-Mark® Anti-CD11c-FITC Antibody, clone KB90

clone KB90, Milli-Mark®, from mouse

Synonym(s):

Integrin alpha-X, Leukocyte adhesion glycoprotein p150,95 alpha chain, Leukocyte adhesion receptor p150,95, Leu M5, CD11, antigen-like family member C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

KB90, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGAX(3687)

General description

CD11c is the 150 kDa (Y-chain of the leucocyte integrin complex P1 50,95. The protein belongs to a family of related heterodimeric proteins designated CD11 in the classification system for human leucocyte differentiation antigens. The CD11 family shares a common 95 kDa li-chain (CD18). In addition to the p150,95 protein, this protein family contains two other members: leucocyte function associated-1 antigen (LFA-1, CD11a) and iC3b receptor (CD11b) (Hogg, 1997). CD11c is expressed strongly on monocytes and weakly on neutrophil granulocytes (Hogg, 1997). Natural killer cells and activated B and T cells also express CD11c (Luk, 1995). Anti-CD11c, KB90, was included in the Sixth International Workshop and Conference on Human Leucocyte Differentiation Antigens, and studies by a number of laboratories confirmed its reactivity with the CD11c antigen (Hogg, 1997).

Specificity

Antibody recognizes Human CD11c antigen.

Immunogen

Hairy cell leukemia cells

Application

Milli-Mark Anti-CD11c-FITC Antibody, clone KB90 is an antibody against CD11c for use in FC.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by flow cytometry using human PBMCs

Target description

150 kDa (Hogg, 1997)

Physical form

Protein A purified
Purified Mouse monoclonal IgG1 conjugated to FITC in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.

Storage and Stability

Maintained refrigerated at 2-8°C from date of receipt. Protected from light in undiluted aliquots for 6 months from date of receipt.

Analysis Note

Control
PBMCs

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service