Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

FCMAB289A4

Sigma-Aldrich

Milli-Mark® Anti-PINK1-Alexa Fluor 488 Antibody, clone N4/49

clone N4/49, Milli-Mark®, from mouse

Synonym(s):

PTEN induced putative kinase 1, protein kinase BRPK, serine/threonine-protein kinase PINK1, PTEN-induced putative kinase protein 1, Parkinson disease (autosomal recessive) 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

N4/49, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PINK1(65018)

Related Categories

General description

Mutations in the PTEN-induced kinase 1 (PINK1) gene located within the PARK6 locus on chromosome 1p35-p36 have recently been identified in patients with recessive early-onset Parkinson disease. Data suggests that PINK1 may be the second most common causative gene next to parkin in parkinsonism with the recessive mode of inheritance.

Specificity

Antibody recognizes human PINK1.

Immunogen

Recombinant protein corresponding to human PINK1.

Application

Milli-Mark Anti-PINK1-Alexa Fluor 488 Antibody, clone N4/49 is an antibody against PINK1-Alexa Fluor 488 for use in FC.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Quality

Evaluated by flow cytometry using HEK293 cells.

Target description

Uniprot describes two isoforms exist with calculated Mw of 63 and 30 kDa

Physical form

Protein G Purified
Purified mouse monoclonal IgG1k conjugated to Alexa Fluor 488 in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
HEK293 cells

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service