M3420V
Anti-Tau-1, clone PC1C6 - 10mg (KC) Antibody

Synonym(s):
Anti-Tau-1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
Recommended Products
biological source
mouse
Quality Level
clone
PC1C6, monoclonal
monoclonal
species reactivity
human
technique(s)
immunohistochemistry: suitable
western blot: suitable
storage temp.
−20°C
Application
Anti-Tau-1 Antibody, clone PC1C6 is an antibody against Tau-1 for use in IH & WB.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service