MAB1789
Anti-Migration Inhibitory Factor Related Proteins 8 and 14 Antibody, clone AHN-17
clone AHN-17, Chemicon®, from mouse
Synonym(s):
MRP-8 & 9, Myeloid-Related Protein-8 & 9, Calgranulin A & B, S100 Calcium-binding Protein A8 & A9, Leukocyte L1 cplx Light & Heavy Chain, Calprotectin
About This Item
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
AHN-17, monoclonal
species reactivity
human
manufacturer/tradename
Chemicon®
technique(s)
immunocytochemistry: suitable
western blot: suitable
isotype
IgG1
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... S100A8(6279)
General description
Specificity
Immunogen
Application
Immunoblotting on un-reduced gels
Optimal dilutions must be determined by the end user.
Inflammation & Immunology
Cytokines & Cytokine Receptors
Linkage
Physical form
Storage and Stability
Other Notes
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service