biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
2E3, monoclonal
mol wt
calculated mol wt 56.12 kDa
observed mol wt ~60 kDa
species reactivity
feline, rat, human, mouse
packaging
antibody small pack of 100 μg
technique(s)
ELISA: suitable
electron microscopy: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable
isotype
IgG1κ
UniProt accession no.
shipped in
dry ice
storage temp.
-10 to -25°C
target post-translational modification
unmodified
Gene Information
rat ... Ina(24503)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in Rat brain stem tissue lysates.
Western Blotting Analysis: A 1:500 dilution of this antibody detected Internexin a in Rat brain stem tissue lysates.
Tested Applications
ELISA Analysis: A representative lot detected Internexin a in ELISA applications (Evans, J., et. al. (2002). J Neurohysiol. 87(2):1076-85).
Immunocytochemistry Analysis: A representative lot detected Internexin a in Immunocytochemistry applications (Yuan, A., et. al. (2006). J Neurosci. 26(39):10006-19; Evans, J., et. al. (2002). J Neurohysiol. 87(2):1076-85).
Electron Microscopy Analysis: A representative lot of this antibody was used for ultrastructural colocalization of Internexin a and NF-M on neurofilament in optic nerve of adult mice. (Yuan, A., et. al. (2006). J Neurosci. 26(39):10006-19).
Western Blotting Analysis: A representative lot detected Internexin a in Western Blotting applications (Yuan, A., et. al. (2006). J Neurosci. 26(39):10006-19; Evans, J., et. al. (2002). J Neurohysiol. 87(2):1076-85 ).
Immunohistochemistry Applications: A representative lot detected Internexin a in Immunohistochemistry applications ( Yuan, A., et. al. (2006). J Neurosci. 26(39):10006-19).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service