Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB858-M

Sigma-Aldrich

RSV GLYCO, MS X X3X-2G (KC)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

antibody form

purified antibody

Quality Level

clone

131-2G
monoclonal

species reactivity

human, mouse

concentration

(Please refer to the Certificate of Analysis for the lot-specific concentration.)

technique(s)

ELISA: suitable
immunofluorescence: suitable

isotype

IgG1

General description

A2

Specificity

Glycoprotein, 89 kD. Recognizes both A and B strains.

Application

Epitope: some type B strains

Physical form

Purified immunoglobulin. 1 mg/mL in 0.02M PB with 0.25M NaCl, pH=7.6 containing 0.1% sodium azide.

Storage and Stability

Maintain at 2 to 8° C in aliquots for up to 12 months after date of receipt. Do not store in diluted format.

Disclaimer

This product is intended for use by IVD manufacturers.
Contact customer service for assistance: oem.reagents@emdmillipore.com
Minimum order quantity required to qualify. Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service