MABC1110
Anti-FGFR4 Antibody, clone BT53
Synonym(s):
CD334, EC: 2.7.10.1, Fibroblast growth factor receptor 4
About This Item
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
BT53, monoclonal
mol wt
calculated mol wt 87.95 kDa
observed mol wt ~N/A kDa
purified by
using protein G
species reactivity
human
packaging
antibody small pack of 100
technique(s)
ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
isotype
IgG2bκ
epitope sequence
Extracellular domain
Protein ID accession no.
UniProt accession no.
storage temp.
-10 to -25°C
Gene Information
human ... FGFR4(2264)
Specificity
Immunogen
Application
Evaluated by Immunocytochemistry in HepG2 cells.
Immunocytochemistry Analysis: A 1:250 dilution of this antibody detected Fibroblast growth factor receptor 4 (FGFR4) in HepG2 cells.
Tested Applications
ELISA Analysis: A representative lot detected Fibroblast growth factor receptor 4 (FGFR4) in ELISA applications.
Inhibition Assay:: A representative lot inhibited growth, replication, and metastasis in RH30 cells.
Immunotoxin: A representative lot of this antibody displayed immunotoxin activity as antibody-drug conjugate with Monomethyl auristatin F or Duocamycin DM.
Flow Cytometry Analysis: A representative lot detected Fibroblast growth factor receptor 4 (FGFR4) in Flow Cytometry applications.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service