MABC945
Anti-TRIM3 Antibody, clone 13D12.1
clone 13D12.1, from mouse
Synonym(s):
Tripartite motif-containing protein 3, Brain-expressed RING finger protein, RING finger protein 22, RING finger protein 97, TRIM3
About This Item
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
13D12.1, monoclonal
species reactivity
rat, human, mouse
technique(s)
immunohistochemistry: suitable
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... TRIM3(10612)
General description
Immunogen
Application
Quality
Western Blotting Analysis: 0.5 µg/mL of this antibody detected TRIM3 in 10 µg of HEK293 cell lysate.
Target description
Physical form
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service