Skip to Content
MilliporeSigma

MABE2184

Sigma-Aldrich

Anti-Islet-1 homeobox Antibody, clone 39.3F7

Synonym(s):

Insulin gene enhancer protein ISL-1

Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

39.3F7, monoclonal

mol wt

calculated mol wt 39.04 kDa
observed mol wt ~35 kDa

purified by

using protein G

species reactivity

rat, mouse, human, chicken

packaging

antibody small pack of 100

technique(s)

electron microscopy: suitable
flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

rat ... Isl1(64444)

Specificity

Clone 39.3F7 is a mouse monoclonal antibody that detects Islet-1 homeobox. It targets an epitope within 172 amino acids from the C-terminal part of the protein.

Immunogen

Recombinant fusion protein corresponding to 172 amino acids from the C-terminal part of rat Islet-1 homeobox protein.

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Islet-1 homeobox in HeLa cell lysate.

Tested Applications

Flow Cytometry Analysis: A representative lot detected Islet-1 homeobox in Flow Cytometry applications (Foo, K.S., et al. (2018). Mol Ther. 26(7):1644-1659).

Immunohistochemistry Analysis: A representative lot detected Islet-1 homeobox in Immunohistochemistry applications (Chai, G., et al. (2014). Nat. Neurosci. 17(9):1171-9; Smith, R., et al. (2017). J. Neurosci. 37(33):7975-7993; Schick, E., et al. (2019). Sci Rep. 9(1):9358).

Western Blotting Analysis: A representative lot detected Islet-1 homeobox in Western Blotting applications (Foo, K.S., et al. (2018). Mol. Ther. 26(7):1644-1659).

Immunoprecipitation Analysis: A representative lot immunoprecipitated Islet-1 homeobox in Immunoprecipitation applications (Velasco, S., et al. (2017). Cell Stem Cell. 20(2):205-217).

Immunofluorescence Analysis: A representative lot detected Islet-1 homeobox in Immunofluorescence applications (Smith, R., et al. (2017). J. Neurosci. 37(33):7975-7993; Foo, K.S., et al. (2018). Mol. Ther. 26(7):1644-1659; Schick, E., et al. (2019). Sci. Rep. 9(1):9358).

Electrophoretic Mobility Shift Assay: A representative lot detected Islet-1 homeobox in Electrophoretic Mobility Shift Assay applications (Du, A., et al. (2009). Diabetes. 58(9):2059-69).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Islet-1 homeobox (UniProt: P61374) also known as Islet-1, is encoded by the Isl1 gene (Gene ID: 64444) in rat. Islet-1 homeobox protein is a transcription factor that plays a crucial role in the development of various tissues, including the heart, motor neurons, and sensory neurons. It is a member of the LIM-homeodomain transcription factor family and is involved in the regulation of gene expression during embryonic development. ISL1 is particularly significant in the development of the heart, where it contributes to the formation of the cardiac outflow tract and the differentiation of pacemaker and conduction cells. In addition to its role in cardiac development, ISL1 is also involved in the specification and differentiation of motor neurons and sensory neurons. Its expression is tightly regulated, and mutations in the ISL1 gene have been associated with congenital heart defects and developmental disorders.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service