MABF1461
Anti-CD62L (L-Selectin) (mouse), redFluor® 710, clone MEL-14 Antibody
clone MEL-14, from rat, redFluor® 710
Synonym(s):
L-selectin, CD62 antigen-like family member L, Leukocyte adhesion molecule 1 (LAM-1), Leukocyte-endothelial cell adhesion molecule 1 (LECAM1), Lymph node homing receptor, Lymphocyte antigen 22 (Ly-22), Lymphocyte surface MEL-14 antigen, CD62L, CD62L (L-S
About This Item
biological source
rat
Quality Level
conjugate
redFluor® 710
antibody form
purified antibody
antibody product type
primary antibodies
clone
MEL-14, monoclonal
species reactivity
mouse
technique(s)
flow cytometry: suitable
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
mouse ... Sell(20343)
General description
Immunogen
Application
Inflammation & Immunology
Immunoglobulins & Immunology
Quality
Flow Cytometry Analysis: 0.5 μg from a representative lot detected CD62L (L-Selectin) in one million C57Bl/6 splenocytes.
Physical form
Storage and Stability
Note: It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.
Other Notes
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service