Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF2165

Sigma-Aldrich

Anti-Influenza A M1/M2 Antibody, clone E10

clone E10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

clone

E10, monoclonal

Quality Level

isotype

IgG2aκ

Specificity

Clone E10 is a mouse monoclonal antibody that detects both M1 and M2 proteins of Influenza A virus.

Immunogen

KLH-conjugated linear peptide corresponding to 11 amino acids from M2 protein of Influenza A virus that protrudes out of the viral and cellular membrane.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service