Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN2638

Sigma-Aldrich

Anti-AIDA1 Antibody, clone 2K9

Synonym(s):

Ankyrin repeat and sterile alpha motif domain-containing protein 1B;Amyloid-beta protein intracellular domain-associated protein 1;AIDA-1;E2A-PBX1-associated protein;EB-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2K9, monoclonal

mol wt

calculated mol wt 139.21 kDa
observed mol wt ~140 kDa

purified by

using protein G

species reactivity

rat

species reactivity (predicted by homology)

canine, porcine, feline, monkey, human

packaging

antibody small pack of 100 μg

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG3κ

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... HIF1AN(55662)

General description

Ankyrin repeat and sterile alpha motif domain-containing protein 1B (UniProt: P0C6S7; also known as Amyloid-beta protein intracellular domain-associated protein 1, AIDA-1, E2A-PBX1-associated protein, EB-1) is encoded by the Anks1b gene (Gene ID: 314721) in rat. AIDA-1 is an adaptor protein that has been implicated in Cajal body and nucleolar function. It is expressed throughout the brain and is highly expressed in the hippocampus. It is one of the most abundant proteins at postsynaptic densities where it is reported to bind to PSD95 and associates with NMDA receptors (NMDARs). In the brain, it interacts with amyloid b precursor protein and plays a role in brain development. AIDA-1 preferentially associates with GluN2B-containing NMDARs and plays a role in their transport from the endoplasmic reticulum (ER). Loss of AIDA-1 results in the accumulation of GluN2B in the ER and decreases GluN2B and increases GluN2A distribution at the synapse. However, loss of AIDA-1 does not affect AMPAR-mediated synaptic transmission or the probability of glutamate release. AIDA-1 has seven Ankyrin repeats in its N-terminal half and has two closely juxtaposed sterile alpha motif (SAM) domains (aa 810-876 and 884-949). Within the second SAM domain resides the nuclear localization signal (aa 935-938). Mutations in Anks1b gene are known to affect GluN2B function and are linked to the pathogenesis of neuropsychiatric disorders. (Ref.: Carbonell, AU., et al., (2019). Nat. Commun. 10(1); 3529; Tindi, JO., et al. (2015). J. Neurosci. 35(24); 8986- 8996).

Specificity

Clone 2K9 is a mouse monoclonal antibody that detects AIDA1/ANKS1B. It targets an epitope within 10 amino acids from the C-terminal half.

Immunogen

A linear peptide corresponding to 10 amino acids from the SAM1/2 domain in the C-terminal half of rat AIDA1/ANKS1B.

Application

Quality Control Testing

Evaluated by Western Blotting in Rat brain tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected AIDA1 in Rat brain tissue lysate (non-reduced).

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected AIDA1 in rat testis tissue lysates (non-reduced).

Immunocytochemistry Analysis: A representative lot detected AIDA1 in Immunocytochemistry applications (Tindi, J.O., et al. (2015). J Neurosci. 35(24):8986-96).

Western Blotting Analysis: A representative lot detected AIDA1 in Western Blotting applications (Tindi, J.O., et al. (2015). J Neurosci. 35(24):8986-96).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-AIDA1, clone 2K9, Cat. No. MABN2638, is a mouse monoclonal antibody that detects AIDA1 and is tested for use in Immunocytochemistry and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG3 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service