Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN2746

Anti-Guanylate cyclase 1 Antibody, clone IS4

new
Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

IS4, monoclonal

species reactivity

mouse, bovine

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

Protein ID accession no.

UniProt accession no.

Gene Information

bovine ... GUCY2D(282245)

Specificity

Clone IS4 is a mouse monoclonal antibody that detects an epitope within 15 amino acids from the C-terminal region of bovine Retinal guanylyl cyclase 1.

Immunogen

Linear peptide corresponding to 15 amino acids from the C-terminal region of bovine Retinal guanylyl cyclase 1.

Target description

Retinal guanylyl cyclase 1 (UniProt: P55203; also known as EC:4.6.1.2, RETGC-1, Guanylate cyclase 2D, retinal, Guanylate cyclase E (GC-E), Rod outer segment membrane guanylate cyclase, ROS-GC) is encoded by the GUCY2D (also known as GUC2D) gene (Gene ID: 282245) in bovine. Guanylate cyclase 1 is a membrane-bound retinal guanylyl cyclase expressed predominantly in the cone and rod photoreceptors in the cone outer segment. Guanylate cyclase 1 and the related Guanylate cyclase 2 mediate the synthesis of cyclic 3 , 5 -guanosine monophosphate (cGMP) from guanosine triphosphate in mammalian photoreceptor cells, where Guanylate cyclase 1 and its associated activator proteins are responsible for the Ca2+-sensitive restoration of cGMP levels after light activation of the photo transduction cascade. In bovine, Guanylate cyclase 1 is produced with a signal peptide (aa 1-56) sequence, the removal of which yields the mature enzyme with an extracellular domain (aa 57-467), followed by a transmembrane segment (aa 468-492) and a cytoplasmic tail (aa 493-1110) that contains a protein kinase homology domain (aa 493-818) and the guanylate cyclase catalytic domain (aa 885-1015). Heterozygous mutations in the GUCY2D gene have been shown to cause autosomal dominant cone and cone-rod dystrophies (adCODs and adCORDs, respectively), while homozygous or compound heterozygous mutations cause autosomal recessively inherited Leber Congenital Amaurosis (LCA), the most severe form of inherited retinopathy, resulting in total blindness or greatly impaired vision at birth or in early infancy. (Ref.:Haire, S.E., et al. (2006). Invest Ophthalmol Vis Sci. 47(9):3745-53; Ying, G., et al. (2018). J Biol Chem. 293(45):17546-17558).

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service