Skip to Content
MilliporeSigma

MABT1442

Sigma-Aldrich

Anti-Podocalyxin Antibody, clone MECA99

Synonym(s):

Podocalyxin-like protein 1;PC;PCLP-1

Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

MECA99, monoclonal

mol wt

calculated mol wt 53.39 kDa
observed mol wt ~140 kDa

purified by

using protein G

species reactivity

mouse

packaging

antibody small pack of 100 μg

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2cκ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... Podxl(27205)

General description

Podocalyxin (UniProt: Q9R0M4; also known as Podocalyxin-like protein 1, PC, PCLP-1) is encoded by the Podxl (also known as Pclp1) gene (Gene ID: 27205) in murine species. Podocalyxin is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-21), which is subsequently cleaved off to produce the mature protein that contains an extracellular domain (aa 22-404), a transmembrane domain (aa 405-425), and a cytoplasmic domain (aa 426-503). Its expression is observed in podocytes, liver cells, hematopoietic progenitor cells, vascular endothelial cells, and a subset of neurons. It is present as a monomer but can oligomerize upon association with the membrane raft. Its O-glycan-rich domain in the extracellular region and the C-terminus PDZ-binding motif in the cytoplasmic tail harbor an apical sorting signal. Its cytoplasmic domain is reported to be essential for the apical membrane targeting and renal tubulogenesis. Its large highly anionic extracellular domain allows to maintain open filtration pathways between neighboring podocyte foot processes. The extracellular domain is also required for microvillus formation. In kidneys, it functions primarily as an anti-adhesive molecule in podocyte epithelial cells, regulating adhesion and cell morphology. In other tissues, it plays subtle roles in tissue remodeling and development. It also participates in a number of cellular functions through its association with the actin cytoskeleton, ezrin, and Na+/H+ exchanger regulatory factor 1 and 2 proteins. It is the most commonly used marker protein for detecting podocytes in urine under conditions such as IgA nephropathy, Henoch-Schönlein purpura, diabetic nephropathy, lupus nephritis, poststreptococcal glomerulonephritis, membranous nephropathy, and focal segmental glomerulosclerosis. Higher expression of podocalyxin is observed in many cancers, especially those that originated from the epithelium and is linked to poor prognosis, more aggressive tumor progression, and chemoresistance. (Ref.: Tran, NL., et al. (2021). Cancers (Basel). 1(12); 2863; Kruse, A., et al. (1999). Biol. Reprod. 61(6); 1393-1401).

Specificity

Clone MECA99 is a rat monoclonal antibody that detects Podocalyxin.

Immunogen

Mouse lymph node stroma.

Application

Quality Control Testing

Evaluated by Western Blotting in bEnd.3 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Podocalyxin in bEnd.3 cell lysate.

Tested applications

Immunofluorescence Analysis: A representative lot detected Podocalyxin in Immunofluorescence applications (Kruse, A., et al. (1999). Biol Reprod. 61(6):1393-401; Brulois, K., et al. (2020). Nat Commun. 11(1):3798).

Immunohistochemistry Applications: A representative lot detected Podocalyxin in Immunohistochemistry applications (Hallmann, R., et al. (1995). Dev Dyn. 202(4):325-32; Kruse, A., et al. (1999). Biol Reprod. 61(6):1393-401).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Physical form

Purified rat monoclonal antibody IgG2c in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service