MABX43
Anti-Marburg Antibody Virus
Synonym(s):
Anti-Marburg virus
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
Recommended Products
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified antibody
clone
monoclonal
form
solution
species reactivity
all
packaging
pkg of 1 mg (MABX43-1KC)
pkg of 10 mg (MABX43-10KC)
technique(s)
ELISA: suitable
isotype
IgG1
relevant disease(s)
infectious diseases
shipped in
wet ice
storage temp.
2-8°C
Specificity
Marburg Virus
Immunogen
Native Marburg Virus
Application
This mouse monoclonal antibody, Cat. No. MABX43, is tested for use in ELISA for the detection of Marburg Virus.
Target description
Marburg virus is considered the most dangerous virus in the world with a fatality ratio of up to 88%. Like Ebola vrus it is an RNA virus in the Filoviridae family. It is a hemorrhagic fever virus that causes convulsions and bleeding of mucous membranes, skin and organs. The reservoir host is the African fruit bat Roesettus aegyptiacus.
Linkage
Both MABX42 and MABX43 recognize Marburg Virus
Physical form
Purified mouse monoclonal in buffer containing 10 mM Phosphate Buffered Saline, pH 7.2 with 0.09% Sodium Azide
Reconstitution
Please refer to Certificate of Analysis for concentration.
Storage and Stability
Store at 2-8 °C. For long-term storage, freeze at -20 °C
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service