Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABX7154-10KC

Sigma-Aldrich

Anti-hu N-term proBNP (NT-proBNP) clone 154

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody form

purified antibody

Quality Level

clone

154
monoclonal

purified by

using Protein A

species reactivity

mouse, human

concentration

(Please refer to the Certificate of Analysis for the lot-specific concentration.)

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG2a

General description

proBNP

Application

Confirmed matched pairing. Capture Antibody MABX7132-1KC; Detector Antibody MABX7154-1KC

Physical form

PBS (pH 7.4) with 0.09% sodium azide

Storage and Stability

Stable for 12 months at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service