PRCYTA-40K-BK48
MILLIPLEX® Non-Human Primate Cytokine/Chemokine/Growth Factor Panel A 48-Plex Premixed Magnetic Bead Panel – BULK (Space Saver) PACKAGING
Synonym(s):
48-Plex Cytokine Beads, Primate Cytokine Panel
About This Item
Recommended Products
species reactivity
nonhuman primates
assay range
accuracy: 80-102%
inter-assay cv: <20%
intra-assay cv: <10%
inter-assay cv: <30%
(TNF-α)
technique(s)
multiplexing: suitable
detection method
fluorometric (Luminex® MAP)
shipped in
wet ice
storage temp.
2-8°C
General description
The MILLIPLEX® Non-Human Primate Cytokine/Chemokine/Growth Factor Panel A 48-Plex Premixed Magnetic Bead Panel is a 48-plex kit to be used for the simultaneous quantification of the following analytes in serum or plasma samples and tissue/cell lysate and culture supernatant samples: BCA-1, sCD137, sCD40L, Eotaxin, sFASL, FGF-2, Fractalkine, G-CSF, GM-CSF, Granzyme A, Granzyme B, IFNa2, IFN?, IL-1a, IL-1ß, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-10, IL-12 (p70), IL-15, IL-16, IL-17A, IL-17E, IL-18, IL-21, IL-22, IL-23, IL-28A, IL-31, IL-33, IP-10, I-TAC, MCP-1, MIG, MIP-1a, MIP-1ß, MIP-3a, Perforin, RANTES, TGFa, TNFa, TNFß, VEGF-A.
Specificity
Application
- Analytes: BCA-1, sCD137, CD40L, Eotaxin, sFASL, FGF-2, Fractalkine, G-CSF, GM-CSF, Granzyme A, Granzyme B, IFNa2, IFN?, IL-1a, IL-1ß, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-10, IL-12 (p70), IL-15, IL-16, IL-17A, IL-17E, IL-18, IL-21, IL-22, IL-23, IL-28A, IL-31, IL-33, IP-10, I-TAC, MCP-1, MIG, MIP-1a, MIP-1ß, MIP-3a, Perforin, RANTES, TGFa, TNFa, TNFß, VEGF-A
- Recommended Sample type: Serum, plasma or tissue/cell lysate and culture supernatant samples
- Recommended Sample dilution: Neat plasma or serum. Tissue culture supernatant may require a dilution with an appropriate control medium prior to assay. RANTES requires a 1:100 dilution of plasma/serum samples.
NOTE: RANTES cannot be plexed with other cytokines in this panel due to a required 1:100 dilution of plasma/serum samples. This premixed panel comes with a RANTES bead, so please plan your samples accordingly.
Note: The serum/plasma samples of some non-human primate species may require up to 1:100 dilution in Assay Buffer for IL-8.
- Assay Run Time: Overnight or two-hour primary incubation. For best results, an overnight incubation is recommended
- Research Category: Inflammation & Immunology
- Research Sub Category: Obesity, Metabolic Disorders, Inflammation & Autoimmune Mechanisms
Features and Benefits
Storage and Stability
- Recommended storage for kit components is 2 - 8°C.
- For long-term storage, freeze reconstituted standards and controls at -20°C. Avoid multiple freeze/thaw cycles.
- DO NOT FREEZE Antibody-Immobilized Beads, Detection Antibody, and Streptavidin-Phycoerythrin.
Legal Information
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2
target_organs
Respiratory Tract
Storage Class
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service