Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S7840

Sigma-Aldrich

CPGWIZ Oct-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

species reactivity

mouse

Quality Level

UniProt accession no.

application(s)

genomic analysis

General description

FUNCTION: SwissProt: Q01860 # Transcription factor that binds to the octamer motif (5′-ATTTGCAT-3′). Prime candidate for an early developmental control gene.
SIZE: 360 amino acids; 38571 Da
SUBCELLULAR LOCATION: Nucleus.
TISSUE SPECIFICITY: Ubiquitous. Expressed at low levels.
SIMILARITY: SwissProt: Q01860 ## Belongs to the POU transcription factor family. Class- 5 subfamily. & Contains 1 homeobox DNA-binding domain. & Contains 1 POU-specific domain.
MISCELLANEOUS: Several pseudogenes of POU5F1 have been described on chromosomes 1, 3, 8, 10 and 12. 2 of them, localized in chromosomes 8 and 10, are transcribed in cancer tissues but not in normal ones and may be involved in the regulation of POU5F1 gene activity in carcinogenesis.

Components

The components of the CPG WIZ Oct-4 Amplification Kit include those required for PCR amplification after bisulfite modification of DNA samples. Sufficient reagents are provided to analyze 25 samples with appropriate controls. U Primer Set7.5 μM each primer (25X) / 35 μL / 2003416 /-15°C to -25°C M Primer Set7.5 μM each primer (25X) / 35 μL / 2003417 / -15°C to -25°C W Primer Set7.5 μM each primer (25X) / 35 μL / 2003511 / -15°C to -25°C Universal 10X PCR Buffer / 265 μL / 90396 / -15°C to -25°C

Legal Information

CPGWIZ is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service