Key Documents
SNAP2IHC2
Millipore
SNAP id® 2.0 Protein Detection System
Double IHC
About This Item
UNSPSC Code:
41116010
eCl@ss:
42029053
NACRES:
NB.23
Product Name
SNAP i.d. 2.0 Protein Detection System-Double IHC,
product line
SNAP id®
technique(s)
immunohistochemistry: suitable (paraffin)
immunohistochemistry: suitable (tissue)
immunohistochemistry: suitable
General description
This system includes the SNAP id® 2.0 Base, two IHC frames and incubation covers, assembly fixtures, vacuum tubing, and a Quick Start Guide.
Application
The SNAP id® 2.0 System for IHC introduces a streamlined, medium through-put IHC capability to our SNAP id® 2.0 family. The frames and slide holders enable you to process up to 24 tissue slides, while our unique vacuum technology evenly remove
Other Notes
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial
Legal Information
SNAP id is a registered trademark of Merck KGaA, Darmstadt, Germany