Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRI-13K

Sigma-Aldrich

Sensitive Rat Insulin RIA

for protein sequence analysis, for protein analysis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

grade

for protein analysis

Quality Level

description

ED<sub>80</sub> = 0.09 ± 0.01 ng/mL ED<sub>50</sub> = 0.22 ± 0.01 ng/mL ED<sub>20</sub> = 0.55 ± 0.01 ng/mL
analytes available: Insulin
0.02-1.0 ng/mL (standard curve range)

feature

accuracy 88–108%

species reactivity

rat

technique(s)

radioimmunoassay: suitable

detection method

radioactive

General description

Insulin, a 5.8 kDa hormone is secreted by the islet beta cells in the pancreas. This kit offers sensitive assay for rat/mouse insulin

Specificity

Rat Insulin I 100% Rat Insulin II 100% Human Insulin 100% Porcine Insulin 100% Sheep Insulin 100% Hamster Insulin 100% Mouse Insulin 100% Rat C-Peptide Not Detectable Glucagon Not Detectable Somatostatin Not Detectable Pancreatic Polypeptide No

Application

Two-day disequilibrium incubation at 4°C. Sample volume 100 μL serum, plasma, or tissue culture media.

Analysis Note

Linearity 84–119%

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - STOT RE 2

target_organs

Respiratory Tract

Storage Class

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service