Key Documents
NICHI-BMTL-LRS
Nichiryo
Nichiryo NichiTip Low Retention, Universal, Sterile Pipette Tips
96 tips/rack, 10 racks/pk, 100-1000ul
Synonym(s):
Sterile tips, Rack tips
About This Item
PDF Icon
Shipped & Sold By Nichiryo America
A Trusted Marketplace Seller
PDF Icon
Have a question?
feature
graduations: Yes
material
polypropylene
volume
1000 μL
General description
NichiTip Universal
  • DNase RNase free and Low Retention (excludes BMT-X and Z series)
  • Universal Fit (fits most pipettes.)
  • Premium Packaging Featuring Stand-Up and Puncture Resistance Bags
  • New Reloading System (reduces plastic waste.)
  • Racked Tips are offered in both "Hinged" and "Lift-off" tops (excludes BMT-X and Z series).
  • 5ML and 10ML tips are designed to fit Nichiryo Pipettes exclusively.
Choose from one of the most recent versions:
Certificates of Analysis (COA)