Key Documents
AX-200-S-R
OMICRON™
OMICRON™ Graduated Universal Pipette Tips
200µl, Sterile, 96/RackX10
Synonym(s):
Filter tip, pipette tip, racked pipette tips
About This Item
PDF Icon
Shipped & Sold By Omicron Scientific
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Omicron Scientific
General description
OMICRON™ Pipette Filter Tips (MicroTips) Product Description
OMICRON™ tips are produced in a fully automatic, human touch free production facility following strict manufacturing norms
These tips are produced from high quality diamond finish mould using special medical grade virgin polypropylene with high transparency
Features
  • Free from DNase, RNase, Human DNA, Pyrogen & PCR inhibitor. Ideal match for high sensitive PCR work
  • Free of Lubricants and heavy metals
  • Profile and dimensions of the tups have been precisely engineered to ensure that the dispensed liquid is predictable and precise for each and every produced tip
  • There are graduation marks on all tips that act as visual checks to minimize any risk of errors during sample aspiration
Legal Information
OMICRON™ is a trademark of Omicron Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)